missing translation for 'onlineSavingsMsg'
Learn More

Zeolite ZSM-5, ammonium, Thermo Scientific Chemicals

45.63 € - 449.24 €

Chemical Identifiers

CAS 1318-02-1
Molecular Formula Al2O5Si
Molecular Weight (g/mol) 162.043
MDL Number MFCD00132601
InChI Key HNPSIPDUKPIQMN-UHFFFAOYSA-N
Synonym kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural
PubChem CID 9942228
IUPAC Name dioxosilane;oxo(oxoalumanyloxy)alumane
SMILES O=[Al]O[Al]=O.O=[Si]=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15412527
SDS View Documents
Thermo Scientific Alfa Aesar
045882.14
25 g
45.63 €
25g
Please to purchase this item. Need a web account? Register with us today!
15422527
SDS View Documents
Thermo Scientific Alfa Aesar
045882.22
100 g
96.88 €
100g
Please to purchase this item. Need a web account? Register with us today!
15432527
SDS View Documents
Thermo Scientific Alfa Aesar
045882.36
500 g
449.24 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support. Used in detergent manufacturing for the applications of the removal of hardness. It is also used as Odor control, industrial absorbents, industrial fillers, gas absorption, soil remediation, wastewater filtration, flocculating agent, animal feeds, hydroponics, molecular sieves, desiccants, lightweight concrete, acoustics and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1318-02-1
162.043
HNPSIPDUKPIQMN-UHFFFAOYSA-N
9942228
O=[Al]O[Al]=O.O=[Si]=O
Al2O5Si
MFCD00132601
kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural
dioxosilane;oxo(oxoalumanyloxy)alumane

Specifications

1318-02-1
Al2O5Si
kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural
HNPSIPDUKPIQMN-UHFFFAOYSA-N
dioxosilane;oxo(oxoalumanyloxy)alumane
9942228
Zeolite ZSM-5
Powder
MFCD00132601
Insoluble in water.
O=[Al]O[Al]=O.O=[Si]=O
162.043
Plastic bottle
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-283-8

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.