Learn More
Zeolite Y, hydrogen, Thermo Scientific Chemicals
Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products. Used in detergent manufacturing for the applications of the removal of hardness.
34.60 € - 406.00 €
Chemical Identifiers
CAS | 1318-02-1 |
---|---|
Molecular Formula | Al2O5Si |
Molecular Weight (g/mol) | 162.043 |
MDL Number | MFCD00132601 |
InChI Key | HNPSIPDUKPIQMN-UHFFFAOYSA-N |
Synonym | kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural |
PubChem CID | 9942228 |
IUPAC Name | dioxosilane;oxo(oxoalumanyloxy)alumane |
SMILES | O=[Al]O[Al]=O.O=[Si]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15422487
|
Thermo Scientific Alfa Aesar
045868.14 |
25 g |
34.60 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15432487
|
Thermo Scientific Alfa Aesar
045868.22 |
100 g |
97.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15442487
|
Thermo Scientific Alfa Aesar
045868.36 |
500 g |
406.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products. Used in detergent manufacturing for the applications of the removal of hardness. It is also used as Odor control, industrial absorbents, industrial fillers, gas absorption, soil remediation, wastewater filtration, flocculating agent, animal feeds, hydroponics, molecular sieves, desiccants, lightweight concrete, acoustics and ceramics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1318-02-1 | |
162.043 | |
HNPSIPDUKPIQMN-UHFFFAOYSA-N | |
9942228 | |
O=[Al]O[Al]=O.O=[Si]=O |
Al2O5Si | |
MFCD00132601 | |
kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural | |
dioxosilane;oxo(oxoalumanyloxy)alumane |
Specifications
1318-02-1 | |
Al2O5Si | |
kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural | |
HNPSIPDUKPIQMN-UHFFFAOYSA-N | |
dioxosilane;oxo(oxoalumanyloxy)alumane | |
9942228 | |
Zeolite Y |
Powder | |
MFCD00132601 | |
Insoluble in water. | |
O=[Al]O[Al]=O.O=[Si]=O | |
162.043 | |
Plastic bottle |
Safety and Handling
missing translation for 'einecsNumber' : 215-283-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only