Learn More
Zeolite Y, ammonium, Thermo Scientific Chemicals
35.20 € - 370.00 €
Chemical Identifiers
CAS | 1318-02-1 |
---|---|
Molecular Formula | Al2O5Si |
Molecular Weight (g/mol) | 162.043 |
MDL Number | MFCD00132601 |
InChI Key | HNPSIPDUKPIQMN-UHFFFAOYSA-N |
Synonym | kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural |
PubChem CID | 9942228 |
IUPAC Name | dioxosilane;oxo(oxoalumanyloxy)alumane |
SMILES | O=[Al]O[Al]=O.O=[Si]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15492477
|
Thermo Scientific Alfa Aesar
045867.14 |
25 g |
35.20 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15402487
|
Thermo Scientific Alfa Aesar
045867.22 |
100 g |
80.40 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15412487
|
Thermo Scientific Alfa Aesar
045867.36 |
500 g |
370.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zeolite Y is a useful metal aluminum silicate for proteomics research. It is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products. It is also used as a molecular sieve and as an adsorbent of a wide variety of cations, such as Na+, K+, Ca2+, Mg2+ and others. Zeolite is used in the development of a variety of bioreactor devices and in the determination of serum triglycerides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products
Chemical Identifiers
1318-02-1 | |
162.043 | |
HNPSIPDUKPIQMN-UHFFFAOYSA-N | |
9942228 | |
O=[Al]O[Al]=O.O=[Si]=O |
Al2O5Si | |
MFCD00132601 | |
kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural | |
dioxosilane;oxo(oxoalumanyloxy)alumane |
Specifications
1318-02-1 | |
Powder | |
MFCD00132601 | |
Insoluble in water. | |
O=[Al]O[Al]=O.O=[Si]=O | |
162.043 | |
Zeolite Y, ammonium |
White | |
Al2O5Si | |
kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural | |
HNPSIPDUKPIQMN-UHFFFAOYSA-N | |
dioxosilane;oxo(oxoalumanyloxy)alumane | |
9942228 |
Safety and Handling
missing translation for 'einecsNumber' : 215-283-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only