missing translation for 'onlineSavingsMsg'
Learn More

Zeolite beta, ammonium, Thermo Scientific Chemicals

45.63 € - 480.39 €

Chemical Identifiers

CAS 1318-02-1
Molecular Formula Al2O5Si
Molecular Weight (g/mol) 162.043
MDL Number MFCD00132601
InChI Key HNPSIPDUKPIQMN-UHFFFAOYSA-N
Synonym kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural
PubChem CID 9942228
IUPAC Name dioxosilane;oxo(oxoalumanyloxy)alumane
SMILES O=[Al]O[Al]=O.O=[Si]=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15472497
View Documents
Thermo Scientific Alfa Aesar
045873.14
25 g
45.63 €
25g
Please to purchase this item. Need a web account? Register with us today!
15482497
View Documents
Thermo Scientific Alfa Aesar
045873.22
100 g
93.67 €
100g
Please to purchase this item. Need a web account? Register with us today!
15492497
View Documents
Thermo Scientific Alfa Aesar
045873.36
500 g
480.39 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions. Used in detergent manufacturing for the applications of the removal of hardness. It is also used as Odor control, industrial absorbents, industrial fillers, gas absorption, soil remediation, wastewater filtration, flocculating agent, animal feeds, hydroponics, molecular sieves, desiccants, lightweight concrete, acoustics and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions (alkylation and acylation)
Specifications

Chemical Identifiers

1318-02-1
162.043
HNPSIPDUKPIQMN-UHFFFAOYSA-N
9942228
O=[Al]O[Al]=O.O=[Si]=O
Al2O5Si
MFCD00132601
kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural
dioxosilane;oxo(oxoalumanyloxy)alumane

Specifications

1318-02-1
Powder
MFCD00132601
Insoluble in water.
O=[Al]O[Al]=O.O=[Si]=O
162.043
Zeolite beta, ammonium
White
Al2O5Si
kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural
HNPSIPDUKPIQMN-UHFFFAOYSA-N
dioxosilane;oxo(oxoalumanyloxy)alumane
9942228
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-283-8

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only