missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals

packaged under argon in glass bottle

117.59 € - 460.29 €

Chemical Identifiers

CAS 7440-64-4
Molecular Formula Yb
Molecular Weight (g/mol) 173.05
MDL Number MFCD00011286
InChI Key NAWDYIZEMPQZHO-UHFFFAOYSA-N
Synonym unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
PubChem CID 23992
ChEBI CHEBI:33381
IUPAC Name ytterbium
SMILES [Yb]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15490957
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
000408.04
2 g
117.59 €
2g
Please to purchase this item. Need a web account? Register with us today!
15400967
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
000408.09
10 g
460.29 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-64-4
173.05
NAWDYIZEMPQZHO-UHFFFAOYSA-N
23992
ytterbium
Yb
MFCD00011286
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
CHEBI:33381
[Yb]

Specifications

7440-64-4
99.9% (REO)
MFCD00011286
NAWDYIZEMPQZHO-UHFFFAOYSA-N
ytterbium
23992
173.04
6.97
Powder
Yb
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
[Yb]
173.05
CHEBI:33381
Air and moisture sensitive
Ytterbium
Videos
Safety and Handling

Safety and Handling

P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c

H228-H260

missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: II

missing translation for 'einecsNumber' : 231-173-2

missing translation for 'rtecsNumber' : ZG1925000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

Product Identifier
  • Ytterbium powder

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H228-Flammable solid.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P233-Keep container tightly closed.
  • P370+P378-In case of fire: Use to extinguish.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.