missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

94.47 € - 1027.11 €

Chemical Identifiers

CAS 1314-37-0
Molecular Formula O3Yb2
Molecular Weight (g/mol) 394.09
MDL Number MFCD00011290
InChI Key UZLYXNNZYFBAQO-UHFFFAOYSA-N
Synonym Ytterbia
IUPAC Name diytterbium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Yb+3].[Yb+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11396218
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011191.14
25 g
94.47 €
25g
Please to purchase this item. Need a web account? Register with us today!
11376218
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011191.22
100 g
241.20 €
100g
Please to purchase this item. Need a web account? Register with us today!
11386218
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011191.A1
1 kg
1027.11 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ytterbium(III) oxide serves as a dopant for silicate mineral crystals in lasers, and a colorant for glasses and enamels. It is employed in the preparation of optical fibers, and special types of alloys. It is used in the preparation of chiral ytterbium complexes which are used as catalysts for enantioselective organic synthesis with high yields. It finds application in optical, electronic and ceramic industries. It is used as additives for fluorescent powder and in the preparation of magnetic bubbles in computers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-37-0
394.09
UZLYXNNZYFBAQO-UHFFFAOYSA-N
diytterbium(3+) trioxidandiide
O3Yb2
MFCD00011290
Ytterbia
[O--].[O--].[O--].[Yb+3].[Yb+3]

Specifications

2227°C
Powder
O3Yb2
Ytterbia
UZLYXNNZYFBAQO-UHFFFAOYSA-N
diytterbium(3+) trioxidandiide
394.08
Ytterbium(III) oxide
1314-37-0
99.9% (REO)
MFCD00011290
Soluble in hot dilute acids. Insoluble in water.
[O--].[O--].[O--].[Yb+3].[Yb+3]
394.09
9.2
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-234-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only