missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

60.20 € - 254.27 €

Chemical Identifiers

CAS 19423-87-1
Molecular Formula Cl3Yb
Molecular Weight (g/mol) 279.40
MDL Number MFCD00149885
InChI Key CKLHRQNQYIJFFX-UHFFFAOYSA-K
Synonym ytterbium iii chloride hydrate, acmc-1bo6s, ytterbium 3+ ion hydrate trichloride, ytterbium 3+ chloride-water 1/3/1
PubChem CID 71317291
SMILES [Cl-].[Cl-].[Cl-].[Yb+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11326228
SDS View Documents
Thermo Scientific Alfa Aesar
011193.09
10 g
60.20 €
10g
Please to purchase this item. Need a web account? Register with us today!
11336228
SDS View Documents
Thermo Scientific Alfa Aesar
011193.18
50 g
254.27 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ytterbium(III) chloride hydrate is used as an effective catalyst to prepare acetals by reacting with trimethyl orthoformate. It reacts with N-alkylsalicylideneimines to prepare yttrium(III) Schiff base complexes. It is widely used as NMR shift reagent, calcium ion probe and in DNA microarrays. Its anhydrous form acts as a Lewis acid and catalyzes the alkylation reactions such as aldol reaction and Pictet-Spengler reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 233-800-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ytterbium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.