missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

66.83 € - 293.46 €

Chemical Identifiers

CAS 15280-58-7
Molecular Formula C6H9O6Yb
Molecular Weight (g/mol) 350.18
MDL Number MFCD00150127
InChI Key OSCVBYCJUSOYPN-UHFFFAOYSA-K
IUPAC Name ytterbium(3+) triacetate
SMILES [Yb+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11352528
SDS View Documents
Thermo Scientific Alfa Aesar
014576.09
10 g
66.83 €
10g
Please to purchase this item. Need a web account? Register with us today!
11342528
SDS View Documents
Thermo Scientific Alfa Aesar
014576.18
50 g
293.46 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Yttrium Acetate is widely applied in electronic ceramics, glass, and electronics. Yttrium compounds are used as a catalyst for ethylene polymerization. Yttrium is used in the production of a large variety of synthetic garnets, and Yttria is used to make Yttrium Iron Garnets, which are very effective microwave filters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15280-58-7
350.18
OSCVBYCJUSOYPN-UHFFFAOYSA-K
[Yb+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
C6H9O6Yb
MFCD00150127
ytterbium(3+) triacetate

Specifications

15280-58-7
99.9% (REO)
MFCD00150127
OSCVBYCJUSOYPN-UHFFFAOYSA-K
ytterbium(3+) triacetate
350.16 (Anhydrous)
Crystalline Aggregates
C6H9O6Yb
Insoluble in water,moderately soluble in strong mineral acids.
[Yb+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
350.18
Ytterbium(III) acetate hydrate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 244-137-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ytterbium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.