Learn More
Tenak™ Yellow Cardboard Cryobox
Cryobox made of water-repellent cardboard for long-lasting durability.
Brand: Tenak™ TE22498
This item is not returnable.
View return policy
Description
- Constructed of fibreboard/cardboard
- Water-repellent for longer durability
- Can be used with a large range of grid dividers
Specifications
Yellow | |
Square | |
136 x 136 x 32 mm |
Cardboard | |
Cryo Box |