missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XRCC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21468-1-AP-20UL
This item is not returnable.
View return policy
Description
XRCC1 (X-ray cross complementing factor-1) is involved in DNA base excision repair. XRCC1 is a 70 kDa protein that has been shown to be physically associated with other DNA repair enzymes, including poly (ADP-ribose) polymerase (PARP), DNA Ligase III and DNA polymerase beta. XRCC1 contains a BRCT domain (for BRCA1 carboxyl terminus) which was originally found in the tumor suppressor protein BRCA1. Recently, the BRCT domain has been shown to mediate the protein-protein interaction of XRCC1 and DNA ligase III-alpha.Specifications
XRCC1 | |
Polyclonal | |
Unconjugated | |
XRCC1 | |
DNA repair protein XRCC1, RCC, XRCC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22594, 7515 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P18887, Q60596 | |
XRCC1 | |
XRCC1 Fusion Protein Ag15172 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |