missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14174-1-AP-150UL
This item is not returnable.
View return policy
Description
Xenotropic and polytropic retrovirus receptor (XPR1) is an Orphan-U GPCR with an unknown ligand. ESTs have been isolated from brain, breast, kidney, pancreas, placenta, prostate, skeletal muscle, testis, and tonsil libraries.Specifications
XPR1 | |
Polyclonal | |
Unconjugated | |
XPR1 | |
Protein SYG1 homolog, SYG1, X receptor, X3, XPR1, XR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19775, 289424, 9213 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UBH6, Q9Z0U0 | |
Xpr1 | |
XPR1 Fusion Protein Ag5373 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |