Learn More
Reagecon X6 Dissolution Media Concentrate -Potassium Chloride 0.05M, 961.5ml of conc. dilutes to 25L
Brand:  Reagecon DBC25-960
X6 Dissolution Media Concentrate -Potassium Chloride 0.05M, 961.5ml of conc. dilutes to 25L
Product Code. 16480644
  • 329.00 € / Pack of 6