Learn More
Mbp Molecular Bio Products X5000 Screw-Cap MCT 2.0ml, PP, Natural, non-knurled, w/o cap, EO-sterile, 500/bag
GREENER_CHOICE
Brand:  Mbp Molecular Bio Products 3470NKES
583.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
X5000 Screw-Cap MCT 2.0ml, PP, Natural, non-knurled, w/o cap, EO-sterile, 500/bag
Product Code. 16436909
  • 662.00 € / Pack of 5000
Estimated Shipment: 04-04-2025