Learn More
Memmert™ X4 Height Adjustable Feet For S160
Brand: Memmert™ S160B29768
This item is not returnable.
View return policy
Specifications
Models U, UF TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 30, 55, 75, 110, 160, 260 | |
Height Adjustable Feet Set |