Learn More
Ansell™ Winter Hyd-Tuf™ 52-590 Series Brown Nitrile Heavy Weight Gloves
Product Code. 12619731
Click to view available options
Size:
9
10
Designed to offer excellent resistance to abrasion and protection against injuries, cuts and punctures.
  • Nitrile impregnated coating
  • Soft, flexible jersey lining
  • Absorbs perspiration and offers close, comfortable fit
  • Cut and sewn construction
  • Cotton jersey liner
  • palm impregnated finish
  • Safety cuff
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile
Certifications/Compliance Standard CATEGORY II EN 388 : 3222, EN 511 : 110
Cuff Material Nitrile
Coating Material Nitrile
Product Type Heavy Weight Gloves
Size 9
Style 52-590
Glove Cuff Style Safety
Lining Cotton Jersey
Palm Type Coated-palm
Quantity 72 Pairs
Resistant To Superior resistance to abrasion and good protection against snags, cuts and puncture
Series 52-590