missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Wilmad Low Profile NMR Tube Rack

Simple but useful, this stainless steel NMR tube rack accommodates 12 tubes with a maximum OD of 5mm.

Brand:  Bel-Art™ 66000-0075

Product Code. 15388648

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Small footprint saves valuable benchtop space

Specifications

Specifications

NMR Tube Rack
Tubes (5 mm)
12
6 x 2
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Wilmad Low Profile NMR Tube Rack > Accommodates 12 tubes with a maximum OD of 5mm.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.