missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ White Disposable Polypropylene Coveralls, 4500 Series
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Height (English):
64 to 67 in.
66 to 69 in.
69 to 71 in.
70 to 74 in.
73 to 76 in.
76 to 78 in.
78 to 81 in.
Height (Metric):
164 to 170 cm
167 to 176 cm
174 to 181 cm
179 to 187 cm
186 to 194 cm
194 to 200 cm
200 to 206 cm
Description
CE Simple, Category 1 coverall is designed for domestic DIY use, light-duty industrial cleaning and maintenance applications, or environments where hygiene and cleanliness are important.
- Excellent economic solution to keep the wearer clean and dry
- Suitable for non-hazardous applications
- Breathable and light-duty protection
- Strong, durable 45gsm polypropylene fabric
- Storm flap over zip
- Elasticated hood, cuffs, ankles, and waist
Specifications
Specifications
Product Type | Protective Coveralls |
Size | Small |
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic |
Hood Type | 2-panel |
Closure Type | Sealed Storm Flap |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'