missing translation for 'onlineSavingsMsg'
Learn More
DuPont™ Proshield™ 20 SFR Product Code.: 12201232
Add to basket

DuPont™ Proshield™ 20 SFR

Product Code. 12201232 Shop All DuPont Products
Large
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy

Product Code. 12201232

Brand: DuPont™ D14591547

Add to basket
Add to basket
This item is not returnable. View return policy

Made from a non-halogenated, flame-retardant polypropylene SMS nonwoven fabric. DuPont™ ProShield™ 20 SFR Hooded Coverall provides limited flame spread protection.

  • Limited flame spread coverall with 3-piece hood
  • Limited flame spread SMS polypropylene nonwoven construction
  • Stitched external orange seams for visual identification and differentiation
  • Elasticated wrists, ankles, face and waist
  • Zipper flap
  • White with orange seams
  • Antistatic and halogen-free.
  • Certified according to Regulation (EU) 2016/425
  • Chemical protective clothing, Category III, Type 5 and 6
  • EN 14116 index 1 (limited flame spread), EN 1073-2 (protection against radioactive contamination)
  • Antistatic treatment (EN 1149-5) - on both sides
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hooded Coveralls
Size Large
Material Anti-static SMS
Color White
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Zipper with Storm Flap
Seam Type Stitched, Taped
Protection Class Category III, Type 5 and 6, EN 1073-2
Certifications/Compliance (EU) 2016/425, EN 14116 index 1, EN 1073-2, EN 1149-5
Description Zipper flap, tunneled cuff and facial elastics
Size (Metric) Chest 100 to 108 cm
For Use With (Application) Petrochemical and railway industries, welding, gas and metal applications
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DuPont™ Proshield™ 20 SFR >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.