missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ White with Blue Panel Combined SMS Laminate Protective Coveralls
141.83 €
Specifications
Product Type | Protective Coveralls |
---|---|
Waist Style | Elastic |
Neck Style | Hooded |
Hood Type | 3-panel |
Protection Class | Category III |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11983784
|
3M™
7100040087 |
Medium | |||||||
Description
Specifications
Protective Coveralls | |
Hooded | |
Category III |
Elastic | |
3-panel | |
Enhanced Gusset |