Learn More
3M™ Disposable Protective Coveralls
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Breathable back panel to help improve air ventilation and reduce heat build-up.
- Knit cuffs provide a soft wrist closure for increased comfort compared to elastic closures.
- Elastic waist and ankles for convenience and freedom of movement.
- Two-way zipper with sealable storm flap for convenience and to help provide additional protection.
Specifications
Specifications
Product Type | Protective Coveralls |
Size | X-Large |
Color | Blue, White |
Waist Style | Elastic |
Cuff Style | Knit |
Neck Style | Hooded |
Protection Class | Category III |