missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Open-Top Caps w/PTFE Faced Silicone Liners for E-Z Ex-Traction™ Vials

Product Code. 10557833 Shop All Wheaton Science Products Products
Click to view available options
Neck Finish:
13-425
15-425
22-400
24-400
No. per Case:
100 Cs.
1000 Cs.
250 Cs.
500 Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

Suitable for storage and containment of volatile solvents, sensitive compounds or corrosive chemicals

  • Autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
No. per Case 500 Cs.
No. per Pack 500/Cs.
Neck Finish 24-400
Certifications/Compliance ISO 9001
Closure Included Yes
Autoclavable Autoclavable
Product Type Open Top Cap
For Use With (Equipment) Glass bottle or vial
Closure Liner PTFE/Silicone
Closure Material Polyethylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.