missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ White Polypropylene Caps with Foam/LDPE Liners
Click to view available options
Neck Finish:
15-415
20-410
24-400
24-410
28-400
28-410
33-400
33-430
Closure Liner:
Foam/LDPE
Polyethylene Foam
Description
- Lined with foam polyethylene
- Excellent barrier to aqueous solutions
- Repeat sealability
Specifications
Specifications
Color | White |
Material | Polypropylene |
No. per Case | 200 Cs. |
Neck Finish | 33-430 |
Certifications/Compliance | ISO 9001 |
For Use With (Equipment) | Glass bottle or vial |
Closure Liner | Foam/LDPE |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'