missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Locking Flange Septa
Provide airtight covers
Brand: DWK Life Sciences 240563
107.30 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Provide airtight covers
- Flange-type stoppers are secured with aluminum crimp seals
- Slotted and three-prong plug stoppers are for freeze-dry applications
Specifications
3.3 cm | |
Autoclavable | |
Chlorobutyl Rubber |
Serum bottles and vials | |
Gray | |
Locking Flange Septa |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction