Learn More
DWK Life Sciences Wheaton™ Stainless-Steel Shaft Assembly Kit For Celstir
Adjustable paddle blade impeller for better mixing
98.25 € - 149.00 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11551295
|
DWK Life Sciences
356880 |
250mL Flask |
106.00 €
Each |
Estimated Shipment: 04-12-2024
Log in to see stock. |
|||||
11511295
|
DWK Life Sciences
356874 |
25mL Flask |
98.25 €
Each |
Estimated Shipment: 04-12-2024
Log in to see stock. |
|||||
11531295
|
DWK Life Sciences
356878 |
125mL Flask |
107.00 €
Each |
Estimated Shipment: 04-12-2024
Log in to see stock. |
|||||
10553062
|
DWK Life Sciences
356885 |
1000mL Flask |
149.00 €
Each |
Estimated Shipment: 04-12-2024
Log in to see stock. |
|||||
11571295
|
DWK Life Sciences
356888 |
6000mL Flask |
138.00 €
Each |
Estimated Shipment: 04-12-2024
Log in to see stock. |
|||||
11581295
|
DWK Life Sciences
356891 |
8000mL Flask |
138.00 €
Each |
Estimated Shipment: 04-12-2024
Log in to see stock. |
|||||
Description
- Provide maximum surface interface between culture and flask atmosphere
- Ideal for microcarrier and suspension cultures such as insect cells, hybridomas, and adapted cell lines
- Made from Wheaton 33 low-extractable borosilicate glass that conforms to ASTM™ Type I, Class A and USP Type I requirements
- Impeller rod does not protrude up through top cap
- Sidearms for 500mL to 8L flask allow vented caps which enables the flask to breathe while protecting from contamination
- 1:1 headspace for good gas exchange
- No contamination from stirring rod
- Bottom dimple on 125mL and larger flask improves circulation and reduces accumulation of cells in center of the flask
- Sizes 125 to 1000mL are graduated in 50mL increments; sizes 3000 to 8000mL are graduated in 500mL increments; sizes 25mL and 50mL are non-graduated
- Completely autoclavable at 121°C for 20 minutes
Order Info
Assemblies 500mL and larger can be used with the vented cap (Cat. No. 22-288-526)
Specifications
Shaft Assembly Kit |