Learn More
DWK Life Sciences Wheaton™ Snap-on Style Stoppers for Vials and Serum Bottles
For maximum sealing of vials with blow-back glass finish
779.00 €
Product Code | Brand | Diameter (Metric) Inner | Material | Hardness | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Material | Hardness | Price | Quantity & Availability | |||
13108720
|
DWK Life Sciences
W224100-342 |
3cm | Bromobutyl Rubber | 50 Durometer |
779.00 €
Pack of 1000 |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||
Description
For maximum sealing of vials with blow-back glass finish
- Made of gray bromobutyl
Not recommended for vials with a straight-walled finish.
Specifications
Autoclavable | |
Gray | |
Plug Stopper |