Learn More
DWK Life Sciences Wheaton™ Micro Tissue Grinder With Cap, PTFE Pestle and Cap Liner
Black phenolic 13-425 open-top screw cap with PTFE-faced silicone rubber liner reduces aerosoling of sample. Unit can be autoclaved or sterilized to prevent cross contamination between samples.
Brand: DWK Life Sciences 357535
This item is not returnable.
View return policy
Description
- Pestle rod made from No. 316 stainless steel rod with pestle tip made from PTFE
- Mortar made from low-extractable borosilicate glass
- Replace open-top screw cap with solid top cap provides vial for temporary sample storage.
Order Info
Clearance between pestle and mortar is 0.05 to 0.15mm. The sample can be stored in mortar by using a Wheaton solid-top black phenolic screw cap with PTFE-faced rubber liner (No. 240408).
Specifications
0.016 oz. | |
Cap, PTFE pestle, cap liner, Borosilicate Glass mortar | |
115mm | |
0.5 mL |
0.016 oz., 0.5mL | |
4.52 in. | |
Autoclavable |