Learn More
DWK Life Sciences Wheaton™ Low-Density Extended Controlled Dropper Tips
Product Code. 12438782 Shop All Wheaton Science Products Products
Click to view available options
Color:
Blue
Gray
Green
Natural
Orange
Pink
Purple
Red
White
Yellow
Material:
LDPE
Polyethylene
8mm extended controlled dropper tip made of LDPE (Low Density Polyethylene)
  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Material LDPE
No. per Case 100 Cs.