missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Low-Density Extended Controlled Dropper Tips

Product Code. 10697333
Click to view available options
Color:
Blue
Gray
Green
Natural
Orange
Pink
Purple
Red
White
Yellow
Material:
LDPE
Polyethylene
This item is not returnable. View return policy
This item is not returnable. View return policy

8mm extended controlled dropper tip made of LDPE (Low Density Polyethylene)

  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 100
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material LDPE
No. per Case 100 Cs.
Certifications/Compliance ISO 9001
Drop Size 41 μL
Product Type Dropper Tip
For Use With (Equipment) Plastic dropping bottle
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'