missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Clear Straight-Sided Jars with TFE-lined Caps
Ideal for soil sampling and environmental applications
63.10 € - 253.00 €
Specifications
Color | Clear |
---|---|
Material | Soda Lime Glass |
Closure Color | White |
Closure Liner | PTFE |
Closure Material | Polypropylene |
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
10632373
|
DWK Life Sciences
W216908 |
60 mL | 53-400 mm |
116.00 €
Pack of 24 |
Estimated Shipment: 11-02-2025
Log in to see stock. |
||||
10317092
|
DWK Life Sciences
W216909 |
125 mL | 58-400 mm |
92.55 €
Pack of 24 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
11591115
|
DWK Life Sciences
W216910 |
250 mL | 70-400 mm |
63.10 €
Pack of 12 |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||
11501125
|
DWK Life Sciences
W216911 |
500 mL | 89-400 mm |
97.75 €
Pack of 12 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
10275302
|
DWK Life Sciences
W216912 |
1000 mL | 89-400 mm |
253.00 €
Pack of 12 |
Estimated Shipment: 03-03-2025
Log in to see stock. |
||||
Description
Ideal for soil sampling and environmental applications
Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass
- Walls allow for complete removal of contents
- Wide-mouth design is great for large solid samples
- Each case of Convenience Pack Bottles includes Hazard Communication labels for proper content identification and Tamper Evident labels to protect sample integrity in transit or long-term storage
Order Info
Available with caps attached or without caps. Caps can be ordered separately.
Compliance
Clear USP Type III soda-lime glass
Specifications
Clear | |
White | |
Polypropylene |
Soda Lime Glass | |
PTFE | |
Screw Cap |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title