missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Black Phenolic Screw Cap with PTFE-Faced Rubber Liner

Product Code. 11774219
Click to view available options
Neck Finish:
13-425
15-415
15-425
18-400
18-415
20-400
28-400
33-400
33-430
No. per Case:
100 Cs.
200 Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

Features PTFE faced (14B) styrene-butadiene rubber liner

  • Designed for product safety
  • TFE/Styrene-Butadiene rubber liner
  • Screw cap
  • Pheolic
  • Black
  • Autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
No. per Case 100 Cs.
Neck Finish 33-400
Certifications/Compliance ISO 9001
Autoclavable Autoclavable
For Use With (Equipment) Bottle
Closure Liner PTFE/14B Rubber
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'