missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Reeve Angel 226 Creped Qualitative Circles
A general qualitative filter with creped surface. High filtration speed, faster than grade 202 and retain coarse particles gelatinous precipitates.
Brand: Cytiva 5226-848
This item is not returnable.
View return policy
Description
- High filtration speed
- Retains coarse or gelatinous particulates
- Unique crêped gray surface provides optical contrast for white or dark filtrates
Specifications
226 8 × 8 in 1000/pk | |
Cellulose | |
20 to 25 μm | |
A general qualitative filter with creped surface. High filtration speed, faster than grade 202 and retain coarse particles gelatinous precipitates. | |
1000 per pack | |
8x8 in. | |
20.3 cm | |
20 cm |
Sheet | |
20 to 25 μm | |
Fast | |
226 | |
0.28 mm | |
8 in. | |
20.3 cm |