Learn More
Cytiva Whatman™ Polycap Disposable Capsules, 75 AS
Polycap 75 AS filters for filtration of aqueous solutions
Brand: Cytiva 6705-7502
This item is not returnable.
View return policy
Description
- Sterile and nonpyrogenic
- Integrity testable
- With inlet vent
- Filter Media: Low-extractable, hydrophilic nylon membrane and glass microfiber prefilter
- Filling bell model (09-744-61) minimizes contamination when filtering into bottle or flask
Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.
Specifications
Autoclavable | |
0.2 μm | |
820 cm2 | |
Polypropylene | |
Stepped Barb |
Nylon | |
Sterile | |
Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris. | |
Stepped Barb |