Learn More
3M™ Welding Visors
Product Code. 11721363
Click to view available options
Product Type:
Welding Visor Headtop
Specifications
Product Type Welding Visor Headtop
For Use With (Application) Molten metal splashes, gas, vapor, and particulates, welding glare, metal fumes, in automotive, welding, and metal fabrication industries
Includes 90 x 110 mm filter holder able to hold fixed shade or auto-darkening filters