missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Weighing PharmaScoop™

Sampling Systems™ Weighing PharmaScoop™ is ideal for use with weighing scales in hygienic environments.

Brand:  Sampling Systems™ A376-500

Product Code. 15501802

  • 99.80 € / Each
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Free standing scoop, ideal for use with weighing scales
  • 316L Stainless Steel
  • Rounded corners for easy cleaning
  • Crevice free construction
  • Satin finish
  • Robust handle
Specifications

Specifications

500 mL
Boat
320 mm
Stainless Steel
Weighing Scoop
110 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers