missing translation for 'onlineSavingsMsg'
Learn More
Water, ultrapure, Spectrophotometric Grade, Thermo Scientific Chemicals Product Code.: 11313557

Water, ultrapure, Spectrophotometric Grade, Thermo Scientific Chemicals

Product Code. 11313557
1 L
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
This item is not returnable. View return policy

Product Code. 11313557

Brand: Thermo Scientific Alfa Aesar 019391.K2

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Maximum level of impurities: Evaporation residue 2ppm, Specific conductance at 25° 1.5μ mhos/cm, SiO2 0.01ppm, Heavy metals (as Pb) 0.01ppm, Substances reducing permanganate P.T.

Water ultrapure is used in spectrophotometry and environmental testing. It is also used in conductometric titration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Melting Point 0°C
Boiling Point 100°C
Solubility Information Miscible with methanol,acetone,ethanol.
Formula Weight 18.02
Grade Spectrophotometric
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Density 1
Quantity 1 L
Refractive Index 1.333
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS29J782 Certificate TypeCertificate of Analysis Date23/06/2023 Catalog Number019391.K2,019391.K7,019391,019391.M1
Lot NumberR30J753 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number019391.K2,019391.K7,019391.M1
Lot NumberN28I747 Certificate TypeCertificate of Analysis Date18/11/2022 Catalog Number019391.K2,019391.0F,019391.K7,019393.K7,019391.M1
Lot NumberJ05U769 Certificate TypeCertificate of Analysis Date14/09/2022 Catalog Number019391.K2,019391.0F,019391.K7,019393.K7,019391.M1
Lot NumberJ20U827 Certificate TypeCertificate of Analysis Date12/07/2022 Catalog Number019391.K2,019391.0F,019391.K7,019393.K7,019391.M1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Water, ultrapure, Spectrophotometric Grade, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.