missing translation for 'onlineSavingsMsg'
Learn More

Water, CHROMASOLV™, for HPLC, Honeywell Riedel-de Haën™

Product Code. 15688350
Click to view available options
Quantity:
1 L
2.5 L
4 L
6 x 1 L
Packaging:
Glass Bottle

CHROMASOLV™, for HPLC

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Absorbance max. 0.005 at 400 nm, max. 0.005 at 250 nm, max. 0.01 at 210 nm, max. 0.01 at 205 nm
Boiling Point 100°C
CAS Max % 100.0000%
Linear Formula H2O
Beilstein 2050024
Formula Weight 18.02
Grade HPLC
Packaging Glass Bottle
Density 1.000 g/mL at 3.98 °C(lit.)
Quantity 4 L
Chemical Name or Material Water, CHROMASOLV™, for HPLC
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'