missing translation for 'onlineSavingsMsg'
Learn More

Water, ACS Reagent, Honeywell Riedel-de Haën™

Product Code. 15648360
4 L, Poly Bottle
Click to view available options
Quantity:
500 mL
2.5 L
4 L
6 x 500 mL
4 x 2 L
4 x 4 L
Packaging:
Fiberboard Box
Glass Bottle
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Melting Point 0°C
Boiling Point 100°C
UN Number NONH for all modes of transport
Beilstein 2050024
Formula Weight 18.02g/mol
Grade ACS Reagent
Packaging Poly Bottle
Density 1.000g/mL at 3.98°C
Quantity 4 L
Chemical Name or Material Water
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Water, ACS Reagent, Honeywell Riedel-de Haën™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.