Learn More
Smeg™ Washer Rack
Designed for use with Smeg™ glassware washing devices. Smeg Washer Racks can be used as racks to hold small glass items or as an alternative to the Nylon holding nets.
Specifications
Material | Stainless Steel |
---|---|
Product Type | Rack |
For Use With (Equipment) | Small glassware, baskets and holding nets |
Description
- Additional accessories for the small glassware baskets CS1-1 and CS2
- Flat AISI 304 stainless steel mesh with small mesh openings
- PF1 bottom rack with hole for positioning on the top level
- PF2 complete bottom rack for positioning on the bottom level only
- PF1/2 half bottom rack for positioning on the top level only
Specifications
Stainless Steel | |
Rack | |
Small glassware, baskets and holding nets |