Learn More
Ohaus™ Wall Mount Clamp
OHAUS™ Wall Mount Clamp provides support when working with labware at elevated positions.
Brand:  Ohaus™ 30392244
Product Code. 15782289
  • 35.90 € / Each
Estimated Shipment: 11-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed to securely grip and position laboratory apparatus
  • Extension arms with a 360° rotation allow placement of apparatus at various distances from lab-frame without compromising experiment integrity
  • Fixed position
  • Dual adjust
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Wall Mount Clamp
Clamp, Specialty, Wall, CLS-WALLCZ
80.00 mm
2-Prong
Nickel-plated
3.15 in.
5 to 10 mm