Learn More
VPS33B Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12195-1-AP-150UL
 View more versions of this product
Product Code. 16869753
  • 410.00 € / 150µL
Explore more special offers
Description
Description
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene is a member of the Sec-1 domain family, and encodes the human ortholog of rat Vps33b which is homologous to the yeast class C Vps33 protein. The mammalian class C Vps proteins are predomitly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway.
Specifications
Specifications
VPS33B
Polyclonal
Unconjugated
VPS33B
hVPS33B, VPS33B
Rabbit
Antigen Affinity Chromatography
RUO
233405, 26276, 541534, 64060
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P59016, Q58EN8, Q63616, Q9H267
vps33b
VPS33B Fusion Protein Ag2833
150 μL
Primary
Human, Mouse, Rat, Zebrafish