Learn More
Sampling Systems™ Volumetric Spoon Set
Sampling Systems™ Volumetric Spoon Set is ideal for measuring liquids or powders at specified volumes.
Brand:  Sampling Systems™ A335-1
Product Code. 15849631
  • 17.90 € / Each
Estimated Shipment: 03-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Crevice free
  • Satin finish
  • 304 Stainless Steel
  • Supplied as a set
  • Each spoon is engraved with volume
Specifications
Specifications
Stainless Steel (AISI 304)
1.25, 5, 15, 100 mL
Polished