Learn More
Hirschmann™ Volumetric Pipets with Attached Syringe
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann™ Volumetric Pipets with Attached Syringe come with amber graduation.
Brand:  Hirschmann™ 1410164
 View more versions of this product
Product Code. 10577441
  • 42.10 € / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • One mark
  • Amber stain graduation
Specifications
Specifications
± 0.03 mL
To Deliver
Soda Lime Glass
Yes
Permanent amber markings with silver diffusion color
± 0.03 mL
Volumetric Pipette Syringe
Autoclavable
Class B
Non-sterile
Amber
1 Each
15 mL
Reusable