Learn More
Buerkle™ Volume Head for PharmaPicker
Volume tips to insert into the PharmaPicker collection cylinder.
Brand:  Buerkle™ 5346-2200
 View more versions of this product
Product Code. 11981561
  • 140.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Alert:
Further sampling volumes available on request!
Description
Description
  • Volume tips to insert into the PharmaPicker collection cylinder
  • Sampling quantity 0.1 - 1.2 ml for collection cylinder 5346-1000
  • Sampling quantity 1.25 - 2.5 ml for collection cylinder 5346-2000
  • Stainless steel AISI 316L (1.4404)
Specifications
Specifications
Volume Head
Stainless Steel (V4A)
2 mL
PharmaPicker Collection Cylinder