missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Vivacon™ Polycarbonate/Polypropylene Centrifugal Unit

148.92 €

Specifications

For Use With (Equipment) PCR
Material Polycarbonate, Polypropylene
For Use With (Application) Concentration, Desalting, buffer exchange
Housing Material Polycarbonate, Polypropylene
Quantity 25/Pk.
View More Specs

Products 2
Product Code Brand MWCO Price Quantity & Availability  
Product Code Brand MWCO Price Quantity & Availability  
11735785
View Documents
Sartorius™
VN01H21ETO
30 kDa This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11745785
View Documents
Sartorius™
VN01H41ETO
100 kDa This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

PCR
Concentration, Desalting, buffer exchange
25/Pk.
Vivacon 500 Centrifugal Unit
Polycarbonate, Polypropylene
Polycarbonate, Polypropylene
DNA
0.1 to 0.5 mL
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.