Learn More
Macherey-Nagel™ Visocolor™ ECO Test Kit Refill Pack
Replace consumed chemicals from test kits. Macherey-Nagel™ Visocolor™ ECO Test Kit Refill Packs are business-priced reagent sets used with ECO test kits. The kits use precisely-dosed single reagents, and results are evaluated visually with high-quality color comparison cards.
Brand: Macherey-Nagel™ 931224
Description
Visocolor™ ECO colorimetric and titrimetric test kits achieve high sensitivity and accuracy by using single reagents that can be dosed precisely and by the ability to compensate for the turbidity and color of water samples. The results are evaluated visually with color comparison cards adjusted to the original colors of standard solutions or photometrically with the PF-3 and PF-12Plus photometers for quantitative evaluation.
Easy
- Chemical analysis without further accessories
- No extensive training necessary
- Color-coded reagents with clear dosing instructions
- Easy to understand instruction manual
Safe
- Pictogram test instructions
- Reaction basis according to international standards
- Compensation of turbidity and color
Unique
- High quality test kits
- Business-prized refill packs available
- Ecologically friendly disposal of used reagents
- Delivered in cardboard box with plastic inlay
Specifications
DEHA | |
Colorimetric | |
N/A | |
Reagents, Instructions | |
1 Each | |
5 mL | |
10 min. |
Yellow to Red | |
DEHA determination | |
0 - 0.01 - 0.03 - 0.05 - 0.10 - 0.15 - 0.20 - 0.25 - 0.30mg/L DEHA | |
125 | |
0.01 to 0.30mg/L | |
Redox reaction | |
Replacement chemicals |