Learn More
Macherey-Nagel™ Visocolor™ Eco Hardness (Total) Test Kit
Provides titrimetric determination of total hardness. Macherey Nagel™ Visocolor™ Eco Hardness (Total) Test Kits are packed in an environmentally friendly box and contain easy to understand instructions in 6 languages.
Brand: Macherey-Nagel™ 931029
This item is not returnable.
View return policy
Description
- In acc. with DIN 38406 E3 and DIN 38409 H6
- Allow enough water to run through copper pipes prior to sampling
- Reagent bottles with clear dosing instructions
Specifications
The total hardness of water is based on its content of alkaline earth ions (calcium and magnesium ions) | |
1 drop = 1 °d | |
ECO Hardness (total) Titrimetric Test Kit |
1 Each | |
110 Tests with an average hardness of 10 °d |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- VISOCOLOR ECO Total Hardness
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : ethanol