missing translation for 'onlineSavingsMsg'
Learn More

Visitors Coats

Product Code. 12971425
Encompass
Click to view available options
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Size X-Large
Material LDPE
Color Blue
Closure Type Hook and Loop
Disposable Single-use
Sterility Sterile
Collar Type Revere
Certifications/Compliance CE
For Use With (Application) Applications: Hygiene rules, food processing, clean rooms
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.