missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Visitors Coats
Click to view available options
Size:
Medium
Large
X-Large
Specifications
Specifications
Size | Medium |
Material | LDPE |
Color | Blue |
Closure Type | Hook and Loop |
Disposable | Single-use |
Sterility | Sterile |
Collar Type | Revere |
Certifications/Compliance | CE |
For Use With (Application) | Applications: Hygiene rules, food processing, clean rooms |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction