Learn More
Thermo Scientific™ VisionMate™ SR Single Rack 2D Barcode Reader
Decode a full rack of barcoded storage tubes in less than ten seconds with the compact, user-friendly Thermo Scientific™ VisionMate™ SR Single Rack 2D Barcode Reader.
Brand:  Thermo Scientific™ 3115-11
Product Code. 11902191
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Decode a full rack of barcoded storage tubes in less than ten seconds with the Thermo Scientific™ VisionMate™ SR Single Rack 2D Barcode Reader. This compact, user-friendly reader provides exceptional flexibility with data export formats, tube type readability, and tube layout options.
  • Controlled via on-screen software interface or remotely via TCP/IP connection
  • Scans a full tube rack in less than ten seconds
  • Simply follow the instructions during software installation and connect the reader to a computer via USB port
  • Integrates easily with LIMS, existing databases or tracking systems
  • Small footprint and compact profile save valuable laboratory space
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VisionMate SR 2-D Bar Code Reader
Software Interface or Remotely via TCP/IP Connection
Barcode Reader
Single Rack
Full tube rack in less than ten seconds
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ VisionMate™ SR Single Rack 2D Barcode Reader > VisionMate SR 2D Barcode Reader