missing translation for 'onlineSavingsMsg'
Learn More

Vinyltrichlorosilane, 97%, Thermo Scientific Chemicals

Product Code. 11351567
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-94-5
Molecular Formula C2H3Cl3Si
Molecular Weight (g/mol) 161.481
MDL Number MFCD00000482
InChI Key GQIUQDDJKHLHTB-UHFFFAOYSA-N
Synonym trichlorovinylsilane, vinyltrichlorosilane, trichloro vinyl silane, vtcs, silane, trichloroethenyl, silane, trichlorovinyl, trichlorovinylsilicon, vinyl trichlorosilane, trichlorovinyl silicane, vinylsilicon trichloride
PubChem CID 6413
IUPAC Name trichloro(ethenyl)silane
SMILES C=C[Si](Cl)(Cl)Cl

Specifications

Boiling Point 91°C to 93°C
Melting Point -95°C
Quantity 500 g
Linear Formula CH2=CHSiCl3
UN Number UN1305
Beilstein 1743440
Flash Point 10°C (50°F)
Formula Weight 161.49
Percent Purity 97%
Refractive Index 1.436
Sensitivity Moisture sensitive
Density 1.27
Chemical Name or Material Vinyltrichlorosilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'